accessibility menu, dialog, popup

UserName

CAS RN 616-24-0

CH3H2NH3C

CAS RN 616-24-0

IUPAC Name: pentan-3-amine
Synonyms: 3-pentanamine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: PQPFFKCJENSZKL-UHFFFAOYSA-N
SMILES: CCC(N)CC
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)