CAS RN 6174-86-3
IUPAC Name:
3-chloro-7-hydroxy-4-methyl-2H-chromen-2-one
Synonyms:
chlorferone chlorferron C10H7ClO3
Molecular Weight (g/mol):
210.61
Molecular Formula:
C10H7ClO3
InChi Key:
ODZHLDRQCZXQFQ-UHFFFAOYSA-N
SMILES:
CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12
1
–
1
of
1
results
1
3-chloro-4-methyl-7-hydroxycoumarin, 97%, Thermo Scientific™
CAS: 6174-86-3 Molecular Formula: C10H7ClO3 Molecular Weight (g/mol): 210.61 MDL Number: MFCD00006851 InChI Key: ODZHLDRQCZXQFQ-UHFFFAOYSA-N Synonym: chlorferone,3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one,3-chloro-4-methyl-7-hydroxycoumarin,chlorferron,3-chloro-7-hydroxy-4-methylcoumarin,7-hydroxy-4-methyl-3-chlorocoumarin,chlorferon,3-chloro-4-methylumbelliferone,3-chloro-7-hydroxy-4-methyl-2-benzopyrone,2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl PubChem CID: 5355079 ChEBI: CHEBI:38620 IUPAC Name: 3-chloro-7-hydroxy-4-methylchromen-2-one SMILES: CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12