accessibility menu, dialog, popup

UserName

OOHNHOO

CAS RN 61935-48-6

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 277.32
Molecular Formula: C15H19NO4
InChi Key: RPJMLWMATNCSIS-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCC1NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)