CAS RN 61941-91-1
IUPAC Name:
5-(3,4-dichlorophenyl)furan-2-carbonyl chloride
Synonyms:
5-(3,4-dichlorophenyl)furan-2-carbonyl chloride
Molecular Weight (g/mol):
275.51
Molecular Formula:
C11H5Cl3O2
InChi Key:
XQRLANCABKJNFO-UHFFFAOYSA-N
SMILES:
ClC(=O)C1=CC=C(O1)C1=CC(Cl)=C(Cl)C=C1
1
–
1
of
1
results
1
5-(3,4-Dichlorophenyl)furan-2-carbonyl chloride, Thermo Scientific™
CAS: 61941-91-1 Molecular Formula: C11H5Cl3O2 Molecular Weight (g/mol): 275.51 MDL Number: MFCD02257993 InChI Key: XQRLANCABKJNFO-UHFFFAOYSA-N Synonym: 5-3,4-dichlorophenyl furan-2-carbonyl chloride,2-furancarbonyl chloride, 5-3,4-dichlorophenyl,5-3,4-dichloro-phenyl-furan-2-carbonyl chloride PubChem CID: 3873501 IUPAC Name: 5-(3,4-dichlorophenyl)furan-2-carbonyl chloride SMILES: ClC(=O)C1=CC=C(O1)C1=CC(Cl)=C(Cl)C=C1