accessibility menu, dialog, popup

UserName

CH3OONH2OSNNa

CAS RN 6209-17-2

IUPAC Name: sodium N-[(4-aminophenyl)(oxido)oxo-λ⁶-sulfanylidene]acetamide hydrate
Synonyms: sodium N-[(4-aminophenyl)(oxido)oxo-λ⁶-sulfanylidene]acetamide hydrate
Molecular Weight (g/mol): 254.24
Molecular Formula: C8H11N2NaO4S
InChi Key: CNTXYOLKLFYMSC-UHFFFAOYSA-M
SMILES: O.[Na+].CC(=O)N=S([O-])(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)