accessibility menu, dialog, popup

UserName

HOOH

CAS RN 623-05-2

IUPAC Name: 4-(hydroxymethyl)phenol
Synonyms: gastrodigenin P-hydroxybenzyl alcohol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: BVJSUAQZOZWCKN-UHFFFAOYSA-N
SMILES: OCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)