CAS RN 623-30-3
IUPAC Name:
(2E)-3-(furan-2-yl)prop-2-enal
Synonyms:
3-(2-furyl)acrylaldehyde
Molecular Weight (g/mol):
122.12
Molecular Formula:
C7H6O2
InChi Key:
VZIRCHXYMBFNFD-HNQUOIGGSA-N
SMILES:
O=C\C=C\C1=CC=CO1
1
–
1
of
1
results
1
3-(2-Furyl)acrolein, 99%
CAS: 623-30-3 Molecular Formula: C7H6O2 Molecular Weight (g/mol): 122.123 MDL Number: MFCD00003256 InChI Key: VZIRCHXYMBFNFD-HNQUOIGGSA-N Synonym: 3-2-furyl acrolein,2-furanacrolein,e-3-furan-2-yl acrylaldehyde,3-furan-2-yl acrylaldehyde,2-propenal, 3-2-furanyl,trans-3-2-furyl acrolein,beta-2-furylacrolein,3-2-furyl acrylaldehyde,unii-ssq86u3drp,furyl acrolein PubChem CID: 1549521 IUPAC Name: (E)-3-(furan-2-yl)prop-2-enal SMILES: C1=COC(=C1)C=CC=O