accessibility menu, dialog, popup

UserName

CAS RN 623-33-6

ClH3COONH3

CAS RN 623-33-6

IUPAC Name: 2-ethoxy-2-oxoethan-1-aminium chloride
Synonyms: glycine, ethyl ester, hydrochloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: TXTWXQXDMWILOF-UHFFFAOYSA-N
SMILES: [Cl-].CCOC(=O)C[NH3+]
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results