CAS RN 626-16-4
IUPAC Name:
1,3-bis(chloromethyl)benzene
Synonyms:
1,3-(bis-chloromethyl)benzene
Molecular Weight (g/mol):
175.05
Molecular Formula:
C8H8Cl2
InChi Key:
GRJWOKACBGZOKT-UHFFFAOYSA-N
SMILES:
ClCC1=CC(CCl)=CC=C1
1
–
1
of
1
results
1
m-Xylylene dichloride, 97%
CAS: 626-16-4 Molecular Formula: C8H8Cl2 Molecular Weight (g/mol): 175.052 MDL Number: MFCD00000912 InChI Key: GRJWOKACBGZOKT-UHFFFAOYSA-N Synonym: 1,3-bis chloromethyl benzene,alpha,alpha'-dichloro-m-xylene,m-xylylene dichloride,benzene, 1,3-bis chloromethyl,m-bis chloromethyl benzene,m-xylylene chloride,m-dichloroxylene,benzene, 1,3-dichloromethyl,ccris 1774,m-xylene, .alpha.,.alpha.'-dichloro PubChem CID: 12275 IUPAC Name: 1,3-bis(chloromethyl)benzene SMILES: C1=CC(=CC(=C1)CCl)CCl