accessibility menu, dialog, popup

UserName

CAS RN 62607-69-6

IUPAC Name: 10-phenyldecan-1-ol
Synonyms: 10-phenyldecan-1-ol
Molecular Weight (g/mol): 234.38
Molecular Formula: C16H26O
InChi Key: WNFXODBOMMSELY-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results