accessibility menu, dialog, popup

UserName

H3CHNNH2O

CAS RN 627-06-5

IUPAC Name: propylurea
Synonyms: propylurea
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: ZQZJKHIIQFPZCS-UHFFFAOYSA-N
SMILES: CCCNC(N)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)