accessibility menu, dialog, popup

UserName

HCCH

CAS RN 628-16-0

IUPAC Name: hexa-1,5-diyne
Synonyms: dipropargyl
Molecular Weight (g/mol): 78.11
Molecular Formula: C6H6
InChi Key: YFIBSNDOVCWPBL-UHFFFAOYSA-N
SMILES: C#CCCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)