accessibility menu, dialog, popup

UserName

ClCl

CAS RN 628-76-2

IUPAC Name: 1,5-dichloropentane
Synonyms: 1,5-dichloropentane
Molecular Weight (g/mol): 141.04
Molecular Formula: C5H10Cl2
InChi Key: LBKDGROORAKTLC-UHFFFAOYSA-N
SMILES: ClCCCCCCl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)