CAS RN 629-08-3
IUPAC Name:
heptanenitrile
Synonyms:
heptanenitrile
Molecular Weight (g/mol):
111.19
Molecular Formula:
C7H13N
InChi Key:
SDAXRHHPNYTELL-UHFFFAOYSA-N
SMILES:
CCCCCCC#N
1
–
1
of
1
results
1
Heptanenitrile, 98%, Thermo Scientific Chemicals
CAS: 629-08-3 Molecular Formula: C7H13N Molecular Weight (g/mol): 111.19 MDL Number: MFCD00039503 InChI Key: SDAXRHHPNYTELL-UHFFFAOYSA-N Synonym: heptanonitrile,hexyl cyanide,enanthonitrile,1-cyanohexane,1-heptanonitrile,heptane-1-nitrile,n-heptanenitrile,unii-ua4z3qt98a,ua4z3qt98a,heptane nitrile PubChem CID: 12372 IUPAC Name: heptanenitrile SMILES: CCCCCCC#N