accessibility menu, dialog, popup

UserName

CAS RN 6291-01-6

CAS RN 6291-01-6

IUPAC Name: hydrogen cyclobutanamine chloride
Synonyms: hydrogen cyclobutylamine chloride
Molecular Weight (g/mol): 107.58
Molecular Formula: C4H10ClN
InChi Key: NFAZOGXQOWEWBM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1CCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results