CAS RN 6292-59-7
IUPAC Name:
4-tert-butylbenzene-1-sulfonamide
Synonyms:
4-tert-butylbenzenesulfonamide
Molecular Weight (g/mol):
213.3
Molecular Formula:
C10H15NO2S
InChi Key:
KYDZEZNYRFJCSA-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(C=C1)S(N)(=O)=O
1
–
3
of
3
results
1
4-tert-Butylbenzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(tert-Butyl)benzenesulfonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
4-tert-Butylbenzenesulfonamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.