accessibility menu, dialog, popup

UserName

CAS RN 63-64-9

CH3H2NOCH3OH3C

CAS RN 63-64-9

IUPAC Name: [(3,4-dimethoxyphenyl)methyl](methyl)azanium
Synonyms: [(3,4-dimethoxyphenyl)methyl](methyl)azanium
Molecular Weight (g/mol): 182.24
Molecular Formula: C10H16NO2
InChi Key: XQODFBIAQVJQHF-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=C(OC)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results