accessibility menu, dialog, popup

UserName

CAS RN 630-06-8

IUPAC Name: hexatriacontane
Synonyms: hexatriacontane
Molecular Weight (g/mol): 506.99
Molecular Formula: C36H74
InChi Key: YDLYQMBWCWFRAI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results