CAS RN 6301-02-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Amino-imidazolidine-2,4-dione, TRC
CAS: 6301-02-6 Molecular Formula: C3 H5 N3 O2 Molecular Weight (g/mol): 115.09 Synonym: 1-Aminohydantoin IUPAC Name: 1-aminoimidazolidine-2,4-dione SMILES: NN1CC(=O)NC1=O