accessibility menu, dialog, popup

UserName

NNH

CAS RN 6312-25-0

IUPAC Name: benzyl[2-(pyridin-2-yl)ethyl]amine
Synonyms: benzyl[2-(pyridin-2-yl)ethyl]amine
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: WUDUNBALAGWKQB-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=N1)NCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results