CAS RN 63149-24-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium inner Salt (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium Inner Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium inner Salt (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium inner Salt (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.