accessibility menu, dialog, popup

UserName

HOON

CAS RN 6318-43-0

IUPAC Name: 3-(pyridin-4-yl)propanoic acid
Synonyms: 3-(pyridin-4-yl)propanoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: WSXGQYDHJZKQQB-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results