accessibility menu, dialog, popup

UserName

ClH3CONH2CH3OH3C

CAS RN 63283-42-1

IUPAC Name: 6,7-dimethoxy-1-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Synonyms: 6,7-dimethoxy-1-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Molecular Weight (g/mol): 243.73
Molecular Formula: C12H18ClNO2
InChi Key: UJXLTDHDLUBZBL-UHFFFAOYNA-N
SMILES: [Cl-].COC1=CC2=C(C=C1OC)C(C)[NH2+]CC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)