accessibility menu, dialog, popup

UserName

CAS RN 63352-99-8

HClH2NNHClCl

CAS RN 63352-99-8

IUPAC Name: hydrogen (3,5-dichlorophenyl)hydrazine chloride
Synonyms: hydrogen (3,5-dichlorophenyl)hydrazine chloride
Molecular Weight (g/mol): 213.49
Molecular Formula: C6H7Cl3N2
InChi Key: BDXZOJVMTJOAPS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results