accessibility menu, dialog, popup

UserName

CAS RN 6341-92-0

ClOOHN

CAS RN 6341-92-0

IUPAC Name: 6-chloro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 6-chloro-1H-indole-2,3-dione
Molecular Weight (g/mol): 181.58
Molecular Formula: C8H4ClNO2
InChi Key: RVXLBLSGEPQBIO-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(NC(=O)C2=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)