CAS RN 6344-61-2
CAS RN 6344-61-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fluoren-1-ol, TRC
CAS: 6344-61-2 Molecular Formula: C13 H10 O Molecular Weight (g/mol): 182.22 Synonym: 9H-Fluoren-1-ol (9CI, ACI),Fluoren-1-ol (6CI, 7CI, 8CI),1-Hydroxyfluorene,NSC 51310 IUPAC Name: 9H-fluoren-1-ol SMILES: Oc1cccc2c1Cc3ccccc23