accessibility menu, dialog, popup

UserName

CAS RN 63503-60-6

IUPAC Name: (3-chlorophenyl)boronic acid
Synonyms: 3-chlorophenylboronic acid
Molecular Weight (g/mol): 156.37
Molecular Formula: C6H6BClO2
InChi Key: SDEAGACSNFSZCU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)