accessibility menu, dialog, popup

UserName

CAS RN 638-67-5

H3CCH3

CAS RN 638-67-5

IUPAC Name: tricosane
Synonyms: tricosane
Molecular Weight (g/mol): 324.64
Molecular Formula: C23H48
InChi Key: FIGVVZUWCLSUEI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)