accessibility menu, dialog, popup

UserName

CH3OOOHNOO

CAS RN 64187-48-0

IUPAC Name: 1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Synonyms: 1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 279.29
Molecular Formula: C14H17NO5
InChi Key: VVKAGQHUUDRPOI-UHFFFAOYNA-N
SMILES: COC(=O)C1CC(O)CN1C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results