CAS RN 64195-85-3
CAS RN 64195-85-3
IUPAC Name:
ethyl 5-methyl-2,4-dioxohexanoate
Synonyms:
ethyl 5-methyl-2,4-dioxohexanoate
Molecular Weight (g/mol):
186.21
Molecular Formula:
C9H14O4
InChi Key:
HBJPUWYQVWIKSD-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)CC(=O)C(C)C
1
–
1
of
1
results
1
Ethyl 2,4-dioxo-5-methylhexanoate, 95%
CAS: 64195-85-3 Molecular Formula: C9H14O4 Molecular Weight (g/mol): 186.21 MDL Number: MFCD07323638 InChI Key: HBJPUWYQVWIKSD-UHFFFAOYSA-N Synonym: ethyl5-methyl-2,4-dioxohexanoate,ethyl 2,4-dioxo-5-methylhexanoate,5-methyl-2,4-dioxohexanoic acid ethyl ester,hexanoic acid,5-methyl-2,4-dioxo-, ethyl ester,5-methyl-2,4-dioxo-hexanoic acid ethyl ester,acmc-1bc7v,ethyl-2,4-dioxo-5-methylhexanoate,ethyl 5-methyl-2,4-dioxo-hexanoate,ethyl 5-methyl-2,4-bis oxidanylidene hexanoate PubChem CID: 3017501 IUPAC Name: ethyl 5-methyl-2,4-dioxohexanoate SMILES: CCOC(=O)C(=O)CC(=O)C(C)C