CAS RN 64302-87-0
CAS RN 64302-87-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R162, MedChemExpress
MedChemExpress R162 is a potent inhibitor of glutamate dehydrogenase 1 (GDH1/GLUD1), with anti-cancer properties.