CAS RN 64451-76-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-[4-(1-Methylpropyl)phenyl]propanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-(p-sec-Butylphenyl)propionic Acid, TRC
CAS: 64451-76-9 Molecular Formula: C13 H18 O2 Molecular Weight (g/mol): 206.28 Synonym: 2-[4-(1-Methylpropyl)-phenyl]propanoic Acid,Ibuprofen Imp. O (EP) IUPAC Name: 2-(4-butan-2-ylphenyl)propanoic acid SMILES: CCC(C)c1ccc(cc1)C(C)C(=O)O