CAS RN 645-49-8
CAS RN 645-49-8
IUPAC Name:
[(1Z)-2-phenylethenyl]benzene
Synonyms:
cis-stilbene cis-diphenylethene isostilbene
Molecular Weight (g/mol):
180.25
Molecular Formula:
C14H12
InChi Key:
PJANXHGTPQOBST-QXMHVHEDSA-N
SMILES:
C(=C\C1=CC=CC=C1)\C1=CC=CC=C1
1
–
1
of
1
results
1
cis-Stilbene, 97%
CAS: 645-49-8 Molecular Formula: C14H12 Molecular Weight (g/mol): 180.25 MDL Number: MFCD00004788 InChI Key: PJANXHGTPQOBST-QXMHVHEDSA-N Synonym: cis-stilbene,z-stilbene,z-1,2-diphenylethene,cis-1,2-diphenylethylene,isostilbene,cis-diphenylethene,stilbene, z,z-2-phenylethenyl benzene,benzene, 1,1'-1,2-ethenediyl bis-, z,z-1,2-diphenylethylene PubChem CID: 5356785 ChEBI: CHEBI:36008 IUPAC Name: (Z)-stilbene SMILES: C1=CC=C(C=C1)C=CC2=CC=CC=C2