accessibility menu, dialog, popup

UserName

CAS RN 64704-23-0

NaNaNH2SSNH2OOOO

CAS RN 64704-23-0

IUPAC Name: disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate
Synonyms: disodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate
Molecular Weight (g/mol): 284.26
Molecular Formula: C6H10N2Na2O4S2
InChi Key: PLVPMKWGXOOSKL-UHFFFAOYNA-L
SMILES: [Na+].[Na+].NC(CSSCC(N)C([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results