CAS RN 64849-39-4
CAS RN 64849-39-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Rubusoside, MedChemExpress
MedChemExpress Rubusoside is a natural sweetener and a solubilizing agent with antiangiogenic and antiallergic properties.