CAS RN 6574-99-8
CAS RN 6574-99-8
IUPAC Name:
3,4-dichlorobenzonitrile
Synonyms:
3,4-dichlorobenzonitrile
Molecular Weight (g/mol):
172.01
Molecular Formula:
C7H3Cl2N
InChi Key:
KUWBYWUSERRVQP-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1Cl)C#N
1
–
2
of
2
results
2
3,4-Dichlorobenzonitrile, 97%, Thermo Scientific™
CAS: 6574-99-8 Molecular Formula: C7H3Cl2N Molecular Weight (g/mol): 172.008 MDL Number: MFCD00016379 InChI Key: KUWBYWUSERRVQP-UHFFFAOYSA-N PubChem CID: 81051 IUPAC Name: 3,4-dichlorobenzonitrile SMILES: C1=CC(=C(C=C1C#N)Cl)Cl