accessibility menu, dialog, popup

UserName

CAS RN 661-95-0

IUPAC Name: 1,2-dibromo-1,1,2,3,3,3-hexafluoropropane
Synonyms: 1,2-dibromo-1,1,2,3,3,3-hexafluoropropane
Molecular Weight (g/mol): 309.83
Molecular Formula: C3Br2F6
InChi Key: KTULQNFKNLFOHL-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(Br)C(F)(F)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results