accessibility menu, dialog, popup

UserName

CAS RN 66304-01-6

OSSOO

CAS RN 66304-01-6

IUPAC Name: 3H-1λ⁶,2-benzodithiole-1,1,3-trione
Synonyms: 1λ⁶,2-benzodithiole-1,1,3-trione
Molecular Weight (g/mol): 200.23
Molecular Formula: C7H4O3S2
InChi Key: JUDOLRSMWHVKGX-UHFFFAOYSA-N
SMILES: O=C1SS(=O)(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results