accessibility menu, dialog, popup

UserName

CAS RN 6635-90-1

H3COCH3NNOO

CAS RN 6635-90-1

IUPAC Name: 2-methoxy-4-methyl-5-nitropyridine
Synonyms: 2-methoxy-4-methyl-5-nitropyridine
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: DJNQRLCFAHKFLZ-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(C=N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results