CAS RN 6639-30-1
CAS RN 6639-30-1
IUPAC Name:
1,2,4-trichloro-5-methylbenzene
Synonyms:
2,4,5-trichlorotoluene
Molecular Weight (g/mol):
195.47
Molecular Formula:
C7H5Cl3
InChi Key:
ZCXHZKNWIYVQNC-UHFFFAOYSA-N
SMILES:
CC1=CC(Cl)=C(Cl)C=C1Cl
1
–
3
of
3
results
1
2,4,5-Trichlorotoluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,4,5-Trichlorotoluene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.