CAS RN 67197-96-0
CAS RN 67197-96-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ (+)-U-50488 hydrochloride
CAS: 67197-96-0 Molecular Formula: C19H26Cl2N2O·HCl