CAS RN 67200-34-4
CAS RN 67200-34-4
IUPAC Name:
13-oxa-10,12,14,16-tetraazatetracyclo[7.7.0.0²,⁷.0¹¹,¹⁵]hexadeca-1(16),2,4,6,9,11,14-heptaen-8-one
Synonyms:
13-oxa-10,12,14,16-tetraazatetracyclo[7.7.0.0²,⁷.0¹¹,¹⁵]hexadeca-1(16),2,4,6,9,11,14-heptaen-8-one
Molecular Weight (g/mol):
224.18
Molecular Formula:
C11H4N4O2
InChi Key:
SFSSAKVWCKFRHE-UHFFFAOYSA-N
SMILES:
O=C1C2=CC=CC=C2C2=NC3=NON=C3N=C12