CAS RN 6722-54-9
CAS RN 6722-54-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(2-Methoxyphenethyl)piperazine, TRC
CAS: 6722-54-9 Molecular Formula: C13H20N2O Molecular Weight (g/mol): 220.32 Synonym: 1-(2-Methoxy-2-Phenylethyl)Piperazine IUPAC Name: 1-(2-methoxy-2-phenylethyl)piperazine SMILES: COC(CN1CCNCC1)c2ccccc2