accessibility menu, dialog, popup

UserName

H3CNHOHOClCH3HBr

CAS RN 67287-95-0

IUPAC Name: 6-chloro-3-methyl-1-(3-methylphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrobromide
Synonyms: skf 83959 hydrobromide
Molecular Weight (g/mol): 398.73
Molecular Formula: C18H21BrClNO2
InChi Key: FHYWNBUFNGHNCP-UHFFFAOYNA-N
SMILES: Br.CN1CCC2=C(Cl)C(O)=C(O)C=C2C(C1)C1=CC(C)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results