accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNH3

CAS RN 68076-36-8

IUPAC Name: tert-butyl N-(4-azaniumylbutyl)carbamate
Synonyms: tert-butyl N-(4-ammoniobutyl)carbamate
Molecular Weight (g/mol): 189.28
Molecular Formula: C9H21N2O2
InChi Key: ZFQWJXFJJZUVPI-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)NCCCC[NH3+]
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)