CAS RN 68087-13-8
CAS RN 68087-13-8
IUPAC Name:
6-(methoxymethyl)-2-(methylsulfanyl)-1,4-dihydropyrimidin-4-one
Synonyms:
6-(methoxymethyl)-2-(methylsulfanyl)-1H-pyrimidin-4-one
Molecular Weight (g/mol):
186.23
Molecular Formula:
C7H10N2O2S
InChi Key:
OWIFCBBLFPQMSJ-UHFFFAOYSA-N
SMILES:
COCC1=CC(=O)N=C(N1)SC
1
–
1
of
1
results
1
4-Hydroxy-6-methoxymethyl-2-(methylthio)pyrimidine, 97%, Thermo Scientific™
CAS: 68087-13-8 Molecular Formula: C7H10N2O2S Molecular Weight (g/mol): 186.229 MDL Number: MFCD00052618 InChI Key: OWIFCBBLFPQMSJ-UHFFFAOYSA-N Synonym: 4-hydroxy-6-methoxymethyl-2-methylthio pyrimidine,6-methoxymethyl-2-methylthio pyrimidin-4 3h-one,6-methoxymethyl-2-methylthio pyrimidin-4-ol,6-methoxymethyl-2-methylsulfanyl pyrimidin-4-ol,maybridge1_006914,6-hydroxy-4-methoxymethyl-2-methylthiopyrimidine,6-methyloxy methyl-2-methylthio-4-pyrimidinol,4 1h-pyrimidinone, 6-methoxymethyl-2-methylthio PubChem CID: 2730176 IUPAC Name: 6-(methoxymethyl)-2-methylsulfanyl-1H-pyrimidin-4-one SMILES: COCC1=CC(=O)N=C(N1)SC