accessibility menu, dialog, popup

UserName

NNNNSOHN

CAS RN 681136-29-8

IUPAC Name: N-phenyl-4-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine-1-carboxamide
Synonyms: N-phenyl-4-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine-1-carboxamide
Molecular Weight (g/mol): 365.46
Molecular Formula: C19H19N5OS
InChi Key: BHBOSTKQCZEAJM-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=CC=C1)N1CCN(CC1)C1=NC(=NS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results