CAS RN 68131-40-8
CAS RN 68131-40-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
5
of
5
results
1
Thermo Scientific Chemicals Tergitol™ 15-S-9
CAS: 68131-40-8 Molecular Formula: [C2H4O]n C13H27O MDL Number: MFCD00132411 SMILES: CCCCCCCC(CCCCC)OCCO-*
2
Polyethylene Glycol Trimethylnonyl Ether (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Polyethylene Glycol Trimethylnonyl Ether (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Polyethylene Glycol Trimethylnonyl Ether (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Thermo Scientific Chemicals TergitolTM 15-S-40, 70% solution in water
CAS: 68131-40-8 Molecular Formula: [C2H4O]n C13H27O MDL Number: MFCD00132411 SMILES: CCCCCCCC(CCCCC)OCCO-*