accessibility menu, dialog, popup

UserName

CAS RN 68399-78-0

OHNNOHSOHOO

CAS RN 68399-78-0

IUPAC Name: 2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Synonyms: heppso
Molecular Weight (g/mol): 268.33
Molecular Formula: C9H20N2O5S
InChi Key: GIZQLVPDAOBAFN-UHFFFAOYNA-N
SMILES: OCCN1CCN(CC(O)CS(O)(=O)=O)CC1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
pH 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)