accessibility menu, dialog, popup

UserName

H3CNCH3Cl

CAS RN 6848-13-1

IUPAC Name: 3-chloro-N,N-dimethylaniline
Synonyms: 3-chloro-N,N-dimethylaniline
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: CHHCCYVOJBBCIY-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results